Order Entry
Singapore
Orders LinkContactUsLinkComponent
Insulating tubes, ceramic

Insulating tubes, ceramic

1,7 mm, 2,7 mm, 200 mm
  ALFA31492.DR
Insulating tubes, ceramic
  ALFA31492.DR
Insulating tubes, ceramic

1,7 mm, 2,7 mm, 200 mm

 
 
   
1 * 200 mm
 

Pure Al₂O₃ Aluminium oxide AL23.

Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.

 
  • Tubing ext.Ø:
    2,7 mm
  • Tubing int.Ø:
    1,7 mm
  • Length:
    200 mm
  • Seq:
    49