NucleoMag Kits for Pathogen Detection
About this item
NucleoMag kits are designed for rapid manual and automated small-scale preparation of viral RNA and DNA and the DNA of microorganisms from various types of clinical samples. NucleoMag® kits allow easy automation on common liquid handling instruments or automated magnetic separators.
- Magnetic bead technology
- NucleoMag® kits for pathogen detection are designed for the isolation of viral RNA and DNA and bacterial DNA from cell-free body fluids such as serum or plasma, blood, saliva, homogenized tissue sample suspensions, stool sample suspensions, and swab washes
Support protocols for automation platforms available on request.
Handling: Magnetic separation.
Hazardous material: No.
Automation support: Compatible with most open robotic platforms.
Caution: Shelf life: 24 months from date of production. For research use only.
2 Options Available Below
Product Details & Documents
NucleoMag kits are designed for rapid manual and automated small-scale preparation of viral RNA and DNA and the DNA of microorganisms from various types of clinical samples. NucleoMag® kits allow easy automation on common liquid handling instruments or automated magnetic separators.
- Magnetic bead technology
- NucleoMag® kits for pathogen detection are designed for the isolation of viral RNA and DNA and bacterial DNA from cell-free body fluids such as serum or plasma, blood, saliva, homogenized tissue sample suspensions, stool sample suspensions, and swab washes
Support protocols for automation platforms available on request.
Handling: Magnetic separation.
Hazardous material: No.
Automation support: Compatible with most open robotic platforms.
Caution: Shelf life: 24 months from date of production. For research use only.
Documents
NucleoMag VET.pdf NucleoMag Virus.pdf
Additional Product Details
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- MANA744200.1
- MANA744200.4
- Target molecule:
- Copurification of DNA and RNA
- Copurification of DNA and RNA
- Refine target molecule:
- Viral RNA and DNA
- Viral RNA and DNA
- Method / Format:
- Magnetic beads
- Magnetic beads
- Prep size:
- Scalable
- Scalable
- Fragment length:
- 300 bp - approx. 50 kbp
- 300 bp - approx. 50 kbp
- Sample size:
- <200 µl whole blood/serum/plasma/swab solution/feces, 10 - 30 mg tissue
- <200 µl whole blood/serum/plasma/swab solution/feces, 10 - 30 mg tissue
- Binding capacity:
- 0,4 µg/µl beads
- 0,4 µg/µl beads
- Operation time:
- 40 - 120 min/96 preps
- 40 - 120 min/96 preps
- Recovery:
- Depending on sample amount and quality
- Depending on sample amount and quality
- Storage temperature:
- 15...25 °C (59...77 °F)
- 15...25 °C (59...77 °F)
Specifications
Product Family Options
Product Information
- FormatContentsVolumeAvailabilityPrice
- MANA744200.1NucleoMag VET kit for viral/bacterial DNA and viral RNA from veterinary samples, 625 g, 96 preps, can be used for PCR, enzymatic reactions, RT-PCR, Isolation of viral and bacterial nucleic acidsNucleoMag VET kit for viral/bacterial DNA and viral RNA from veterinary samples, 625 g, 96 preps, can be used for PCR, enzymatic reactions, RT-PCR, Isolation of viral and bacterial nucleic acidsMagnetic beadsAnimal tissue, Plasma, Serum, Blood, Stool, Swabs, Cell-free biological fluid50 - 100 µlLog in for availability1 * 96 TestsSpecifications:
More
Specifications
Cat. No.MANA744200.1Target moleculeCopurification of DNA and RNARefine target moleculeViral RNA and DNAMethod / FormatMagnetic beadsPrep sizeScalableFragment length300 bp - approx. 50 kbpSample size<200 µl whole blood/serum/plasma/swab solution/feces, 10 - 30 mg tissueBinding capacity0,4 µg/µl beadsOperation time40 - 120 min/96 prepsRecoveryDepending on sample amount and qualityStorage temperature15...25 °C (59...77 °F) - MANA744200.4NucleoMag VET kit for viral/bacterial DNA and viral RNA from veterinary samples, 2032,7 g, 384 preps, can be used for PCR, enzymatic reactions, RT-PCR, Isolation of viral and bacterial nucleic acidsNucleoMag VET kit for viral/bacterial DNA and viral RNA from veterinary samples, 2032,7 g, 384 preps, can be used for PCR, enzymatic reactions, RT-PCR, Isolation of viral and bacterial nucleic acidsMagnetic beadsAnimal tissue, Plasma, Serum, Blood, Stool, Swabs, Cell-free biological fluid50 - 100 µlLog in for availability1 * 384 TestsSpecifications:
More
Specifications
Cat. No.MANA744200.4Target moleculeCopurification of DNA and RNARefine target moleculeViral RNA and DNAMethod / FormatMagnetic beadsPrep sizeScalableFragment length300 bp - approx. 50 kbpSample size<200 µl whole blood/serum/plasma/swab solution/feces, 10 - 30 mg tissueBinding capacity0,4 µg/µl beadsOperation time40 - 120 min/96 prepsRecoveryDepending on sample amount and qualityStorage temperature15...25 °C (59...77 °F)



