Order Entry
Singapore
ContactUsLinkComponent
Hype-Wipe® and Mini Hype-Wipe® Disinfecting Towels with Bleach
Hype-Wipe® and Mini Hype-Wipe® Disinfecting Towels with Bleach
Catalog # VWRMNP9803
Supplier:  VWR Products
New Product
undefined
Hype-Wipe® and Mini Hype-Wipe® Disinfecting Towels with Bleach
Catalog # VWRMNP9803
Supplier:  VWR Products
New Product
Specifications
  • Description:
    Mini towels
  • L×W:
    76×76 mm
Specifications
About this item
HYPE-WIPE® Bleach Towelettes are individually packaged, hospital-grade disinfecting/cleaning towelettes saturated with sodium hypochlorite (bleach) diluted to 0,525%, the 1:10 dilution of bleach recommended by the CDC, OSHA, and NCCLS.
Each HYPE-WIPE® consists of a pocket-sized, foil-lined pouch containing a towelette that unfolds to a 152,4×304,8 mm size with enough pre-diluted solution to wipe down a 40 sq. ft. area. MINI HYPE-WIPE® Bleach Towelettes unfold to 76,2×76,2 mm size. The bleach solution is stabilised for a 12-month shelf life with the expiration date on each pouch and on the outside of each box.
HYPE-WIPES® are used to clean and disinfect countertops and laboratory equipment, including centrifuges, analysers, molecular and DNA testing equipment, and slide prep. Mini towelettes are ideal for cleaning up small spills. Safe for use on hard, non-porous surfaces including plastics, Plexiglas®, glass, glazed ceramic tile, linoleum, baked enamel. For stainless steel: follow equipment manufacturer's directions which may specify that surface be rinsed after disinfection. Note: Solution will corrode aluminium.
Note: Solution will corrode aluminium.