NANOCOLOR® Photometric Tube Test Kit for POC
Catalog # MANA985070
Supplier: MACHEREY-NAGEL
CAS Number:
Specifications
- Chemical method:Turbidity
- Pk:1 KIT
- Description:NANOCOLOR® POC 200, test no. 0-70
- Dimensions:160×110×119 mm (Including packaging)
- Platform:NANOCOLOR® tube tests
- Storage temperature:15...25 °C
- Shelf life (months):18
- Weight:438,5 g (Including packaging)
- For unit:UV/VIS II; VIS II; 500 D; PF-12Plus; Advance
- Tests per kit:20
- Range:20 - 200 mg/L (POC AS 2020); 20 - 200 mg/L (POC HS 2020)
Specifications
About this item
Photometric test kit for the determination of POC in water samples.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
- Precisely pre-dosed reagents with constant high quality from batch to batch
- Coloured pictogram instructions and easy handling of the tubes
- 16 mm (OD) tubes for easy pipetting
The test kit is evaluated in 16 mm (AD) round cells. It is pre-programmed on the NANOCOLOR® photometers. Programming data for other spectrophotometers are available.
Sea water analysis: Yes
Remark: Measuring range on NANOCOLOR VIS II
Hazardous material: No
Sludge reagent set: No
Digestion set: No
NanOx Metal: No
NanOx N: No
Delivery:
Rugged box with 20 test tubes. Sufficient for 20 tests.