Order Entry
Singapore
ContactUsLinkComponent
NANOCOLOR® Photometric Standard Tests, Manganese
NANOCOLOR® Photometric Standard Tests, Manganese
Catalog # MANA91860
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
NANOCOLOR® Photometric Standard Tests, Manganese
Catalog # MANA91860
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    Formaldoxime
  • Pk:
    1 KIT
  • Description:
    NANOCOLOR® Manganese , test no. 1-60
  • Dimensions:
    160×110×130 mm (Including packaging)
  • Seq:
    1
  • Model:
    MANA91860
  • Platform:
    NANOCOLOR® standard tests
  • Storage temperature:
    15...25 °C
  • Shelf life (months):
    36
  • Weight:
    421 g (Including packaging)
  • For unit:
    UV/VIS II;VIS II;Advance;500 D
  • Tests per kit:
    250
  • Range:
    0,01 - 10,0 mg/L (Mn)

Specifications

About this item

Photometric test kits for the determination of manganese in water samples.

  • Ready-to-use reagents for up to 500 determinations
  • Excellent sensitivity and accuracy due to measurements in 50 mm cuvettes
  • Low costs per determination
  • Easy dilutions for analysis of higher concentrations

The test kits are evaluated in 10 or 50 mm rectangular cuvettes. They are pre-programmed on the NANOCOLOR® photometers. Programming data for other spectrophotometers are available.

Sea water analysis: No

Hazardous material: Yes

Remark: Measuring range on NANOCOLOR VIS II

Sludge reagent: No

Digestion: No

NanOx metal: No

NanOx: No

Delivery:

Rugged box with reagents for 250 tests.