NANOCOLOR® Photometric Standard Tests Iron
MANA91836
:
- Chemical method:1,10–Phenanthroline
- Pk:1 KIT
- Description:NANOCOLOR® Iron, test no.1-36
- Dimensions:160×110×130 mm (including packaging)
- Seq:1
- Model:MANA91836
- Storage temperature:15…25 °C
- Shelf life (months):36
- Weight:463,3 g (including packaging)
- Type:Sea water compatible, Hazardous material: Yes
- For unit:UV/VIS II; VIS II; Advance; 500 D; MT Easy VIS; MT Easy UV
- Tests per kit:250
- Range:0,01 - 15,0 mg/L (Fe)
Photometric test kits for the determination of iron in water samples.
- Ready-to-use reagents for up to 500 determinations
- Excellent sensitivity and accuracy due to measurements in 50 mm cuvettes
- Low costs per determination
- Easy dilutions for analysis of higher concentrations
The test kits are evaluated in 10 or 50 mm rectangular cuvettes. They are pre-programmed on NANOCOLOR® photometers and are also compatible with METTLER TOLEDO easy VIS and easy UV. Programming data for other spectrophotometers are available.
Sludge reagent set: No
Digestion set: Yes
NanOx Metal: Yes
NanOx N: No
: Remark: Measuring range on NANOCOLOR VIS II