Order Entry
Singapore
ContactUsLinkComponent
Anti-IgG Goat Polyclonal Antibody [clone: ] (Alexa Fluor® 488)
Anti-IgG Goat Polyclonal Antibody [clone: ] (Alexa Fluor® 488)
Catalog # ABCAAB150113-500
Supplier:  ABCAM
undefined
Anti-IgG Goat Polyclonal Antibody [clone: ] (Alexa Fluor® 488)
Catalog # ABCAAB150113-500
Supplier:  ABCAM

Specifications

  • Size:
    500 µg
  • Pack type:
    Vial

Specifications

About this item

Goat Anti-Mouse IgG H&L (Alexa Fluor® 488).

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse