Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-L-phenylalanine
3-Bromo-L-phenylalanine 95%
3-Bromo-L-phenylalanine 95%
3-Bromo-L-phenylalanine 95%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
3-Bromo-L-phenylalanine 95%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
82311-69-1
:
Formula:
C₉H₁₀BrNO₂
MDL Number:
MFCD06659110
:
244.1 g/mol
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH52114
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation