Order Entry
Singapore
Orders LinkContactUsLinkComponent
Absorbent papers
Supplier:   MACHEREY-NAGEL
Absorbent papers
Supplier:  MACHEREY-NAGEL
Certificates
Absorbent papers
Absorbent papers
5 Options Available Below
Product Details & Documents
Filter paper coated on one side with polyethylene for covering laboratory work benches absorbs spilt liquids.
Suitable for isotope and bacteriological laboratories as well as for chemical storerooms and cupboards.
Product Family Options
Product Information
  • L×W
    Thickness
    Availability
    Price
  • MANA112050
    Absorbent paper roll
    Absorbent paper roll
    480×500 mm
    0,22 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll
  • MANA112000.1
    Absorbent paper sheets
    Absorbent paper sheets
    480×600 mm
    0,22 mm
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
  • MANA112010
    Absorbent paper roll
    Absorbent paper roll
    480×1 000 mm
    0,22 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll
  • MANA112030
    Absorbent paper roll
    Absorbent paper roll
    600×500 mm
    0,22 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll
  • MANA112020
    Absorbent paper roll
    Absorbent paper roll
    600×1 000 mm
    0,22 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll