4-(4-Chlorophenoxy)aniline 97%, rods
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:101-79-1
:
Formula:C₁₂H₁₀ClNO
MDL Number:MFCD00043925
Melting Pt:100…103 °C
:219.67 g/mol
EINECS:202-976-5
:GHS08
:GHS07
Storage Temperature:Ambient