Progesterone
Glass bottle for solids, 120 mg
Catalog # EDQMP3300000
Supplier: EDQM
Jump to:
Product Details & Documents
- CAS Number:57-83-0
- Formula:C₂₁H₃₀O₂
- MDL Number:MFCD00003658
- EINECS:200-350-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:120 mg



