2,4-Difluoro-5-nitrophenol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:113512-57-5
:
Formula:C₆H₃F₂NO₃
MDL Number:MFCD08443942
:175.09 g/mol
Melting Pt:112…120 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,II
:GHS07