
Neodymium(III) sulphate octahydrate ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13477-91-3
:
Formula:Nd₂S₃O₁₂·8H₂O
MDL Number:MFCD00149803
:720.78 g/mol (576.66 anhy)
Merck Index:14,6450
Melting Pt:1176 °C
Density:2,85
EINECS:233-262-1
:GHS07
Storage Temperature:Ambient