

Acetone cyanohydrin stabilised
Catalog # PFALA04460-500G
Supplier: PFALTZ BAUER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:85.11 g/mol
EINECS:200-909-4
UN:1541
ADR:6.1,I
:GHS09
:GHS06
CAS Number:75-86-5
Formula:C₄H₇NO
MDL Number:MFCD00004455