Order Entry
Singapore
ContactUsLinkComponent
4-Oxocyclohexanecarboxylic acid ≥98%
Irritant
4-Oxocyclohexanecarboxylic acid ≥98%
Catalog # ALFAH27294.03
CAS Number:  
Irritant
4-Oxocyclohexanecarboxylic acid ≥98%
Catalog # ALFAH27294.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:874-61-3
 :
Formula:C₇H₁₀O₃
 :142.16 g/mol
Melting Pt:65…71 °C
 :GHS07
Storage Temperature:Ambient
MDL Number:MFCD00102035