ExoSAP-IT® reagent for PCR product clean up
ExoSAP-IT® reagent is designed for simple, quick PCR clean up for downstream applications, such as DNA sequencing or Single Nucleotide Polymorphism (SNP) analysis. When PCR amplification is complete, any unconsumed dNTPs and primers remaining in the PCR product mixture will interfere with these methods. ExoSAP-IT® removes these contaminants.
- Add ExoSAP-IT® directly to PCR product for one tube/one step PCR clean-up
- Eliminates spin columns, decreasing time and expense while increasing yield
- 100% recovery of both short and long PCR products conserves PCR samples
- Removes contaminating primers and dNTPs, which interfere in downstream applications
- Generate less waste with a single tube solution to PCR purification
- Method / Format:Enzymatic clean-up
- Prep size:Scalable
- Refine target molecule:Amplified/modified DNA
- Sample type:PCR and other enzymatic reactions
- Target molecule:DNA
- Description:ExoSAP-IT®