Monoparameter photometers, Lovibond® MD 100
: Lovibond
The MD 100 range of photometers are portable handheld instruments with a simple button layout, robust waterproof design and large backlit display that are ideal for testing in the field or laboratory conditions.
- Accurate (3% FS), reproducible results using high quality interference filters with long-life LEDs
- Internal ring memory for 16 data sets
- Real time clock and date
- Optional infrared interface for transfer of test data to PC or printer
: Instrument supplied with carry case, 4× AAA batteries, 3× round glass vials with lids, relevant reagents, and instruction manual.
- Model:MD 100
- Automatic switch-off:Yes
- Keypad:4 button membrane keypad
- Optics:LED, interference filters (IF) and photo sensor in transparent sample chamber
- Power supply:4× AAA batteries
- W×D×H:155×75×35 mm