
2-Amino-4,4,4-trifluorobutyric acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15959-93-0
:MFCD00671488
Formula:C₄H₆F₃NO₂
MDL Number:MFCD00041417
:157.09 g/mol
Melting Pt:∼ 250 °C (dec.)
:GHS07
Storage Temperature:Ambient