Order Entry
Singapore
ContactUsLinkComponent
1,1,2-Trichlorotrifluoroethane
Environment
1,1,2-Trichlorotrifluoroethane
Catalog # EDQMT1985000
Supplier:  EDQM
CAS Number:  
Environment
1,1,2-Trichlorotrifluoroethane
Catalog # EDQMT1985000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 mL
  • Seq:
    2175

Specifications

About this item

 :
 :187.38 g/mol
Boiling Pt:47,6 °C (1013 hPa)
Melting Pt:–35 °C
Density:1,5731 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-936-1
UN:3082
ADR:9,III
 :GHS09
CAS Number:76-13-1
Formula:Cl₂FCCClF₂