
Barium acetate 99.0-102.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
BAKR0942-01
: Avantor
:
- Pack type:Poly bottle, white, blue T/E cap
- Pk:500 g
- Seq:3645
- Meets ACS Reagent Chemical Requirements:99.0 - 102.0%
- For Laboratory,Research,or Manufacturing Use:≤0.01%
- Assay ((CH₃COO)₂Ba):99.0 - 102.0 %
- Insoluble Matter:≤0.01 %
- pH of 5% Solution at 25°C:7.0 - 9.0
- Chloride (Cl):≤0.001 %
- Oxidizing Substances (as NO₃):≤0.005 %
- Calcium (Ca):≤0.05 %
- Potassium (K):≤0.003 %
- Sodium (Na):≤0.005 %
- Strontium (Sr):≤0.2 %
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
- Trace Impurities - Iron (Fe):≤10 ppm
:
:255.42 g/mol
:13,00968
:∼450 °C
:2,47 g/cm³ (20 °C)
:Ambient
:208-849-0
:1564
:6.1,III
:GHS07
:543-80-6
:(H₃CCOO)₂Ba
:MFCD00012447