

Lithium cyanide
Catalog # PFALL03950.10G
Supplier: PFALTZ BAUER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:32.96 g/mol
Storage Temperature:Ambient
EINECS:219-308-3
UN:2265
ADR:3,III
:GHS06
:GHS09
CAS Number:2408-36-8
Formula:CLiN