Ultra Adhesive Labels
Supplier: Sampling Systems
Certificates
About this item
Ultra Adhesive (UA) Labels are special high quality labels with an ultra-adhesive foil backing.
- High strength adhesive - will even stick on very dusty surfaces
- Working temperature range –20 to +70 °C
- Dust and weatherproof
UA Labels are ideal for sealing sacks after sampling. The label will not only record the fact that the sack has been sampled but it will prevent the sack from leaking powder as it is handled.