Order Entry
Singapore
ContactUsLinkComponent
Pepstatin A, MP Biomedicals
Pepstatin A, MP Biomedicals
Supplier:  MP Biomedicals
Certificates
Pepstatin A, MP Biomedicals
Supplier:  MP Biomedicals

Specifications

  • CAS No.:
    26305-03-3
  • Storage conditions:
    Store at −0 °C, protect from light
  • Form:
    White powder
  • MDL No.:
    MFCD00060740
  • Protein/peptide name:
    Pepstatin A
  • Molecular weight:
    685,91 kDa
  • Molecular formula:
    C₃₄H₆₃N₅O₉

Specifications