Cyclosporin A
Catalog # EDQMY0000361
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg
- Storage conditions:Refrigerator
- Protein/peptide name:Cyclosporin A
Specifications
About this item
Signal word: Danger
GHS Symbols: GHS08, GHS07