Order Entry
Singapore
ContactUsLinkComponent
Cresol red sodium salt 0,04% in aqueous solution
Cresol red sodium salt 0,04% in aqueous solution
Catalog # RICC2450-16
Supplier:  Ricca Chemical
CAS Number:  
Cresol red sodium salt 0,04% in aqueous solution
Catalog # RICC2450-16
Supplier:  Ricca Chemical
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :404.42 g/mol
EINECS:263-654-8
CAS Number:62625-29-0
Formula:C₂₁H₁₇NaO₅S
MDL Number:MFCD00001618