VWR®, Bovine deoxyribonuclease I (from Pancreas)
VWRC0649-100KU
:
- Pk:100 KU
- CAS No.:9003-98-9
- Source:Pancreas
- Species:Bovine
- Seq:02
- Storage conditions:Freezer
- Form:Lyophilized powder
- Enzyme name:Deoxyribonuclease I
- Enzyme activity:1800 u/mg (Kunitz, Protein at 25 °C)
- Enzyme synonyms:Dnase I
- Grade:Ultrapure
Deoxyribonuclease I efficiently hydrolyses single-stranded or double-stranded DNA in the presence of divalent cations. Useful in RNA preparations from tissue or bacterial cell cultures, DNase I is a chromatographically purified preparation supplied as a lyophilised powder.
- RNase-free