Kanamycin sulphate hydrate, min. 750 I.U./mg (dried basis) Ph. Eur., BP, pharma grade
Supplier: PanReac AppliChem
Kanamycin sulphate hydrate, min. 750 I.U./mg (dried basis) Ph. Eur., BP, pharma grade
Supplier: PanReac AppliChem
Certificates
About this item
- CAS Number:5965-95-7
- Formula:C₁₈H₃₆N₄O₁₁·H₂SO₄·H₂O
- Molecular Weight:600.60 g/mol
- Storage Temperature:Ambient
- EINECS:246-933-9
3 Options Available Below
Product Details & Documents
- CAS Number:5965-95-7
- Formula:C₁₈H₃₆N₄O₁₁·H₂SO₄·H₂O
- Molecular Weight:600.60 g/mol
- Storage Temperature:Ambient
- EINECS:246-933-9
Documents
Specifications for Product Family
Specification Test Results
- Assay (dried subst.):min. 750 I.U./mg
- α20°C/D; 1 %, H2O, dried subst.:+112° - +123°
- pH (1 %; H2O; 25°C):6.5 - 8.5
- Loss on drying (0.67 kPa; 3 h; 60°C):max. 1.5 %
- Sulfated ash:max. 0.5 %
- Identity:passes test
- Kanamycin B:max. 4.0 %
- Sulfate (dried subst.):15.0 - 17.0 %



