VWR®, SOC medium, in tubes, biotechnology grade
Catalog # VWRCN549-6X5ML
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pk:1 KIT
- Description:SOC outgrowth media
- Volume:6×5 ml
Specifications
About this item
SOC medium is used in the final step of bacterial cell transformation to obtain maximum transformation efficiency of E. coli SOC.
- Ready to use and premixed
- Allows expression of antibiotic resistance genes
Formulation: Tryptone/casein peptone 2,0%, yeast extract 0,5%, sodium chloride 8,5 mM, potassium chloride 2,5 mM, D-glucose 20,0 mM, magnesium chloride hexahydrate 10,0 mM, magnesium sulfate heptahydrate 10,0 mM.