Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Potassium nitrate
Potassium nitrate 100 g/l in aqueous solution
Potassium nitrate 100 g/l in aqueous solution
Potassium nitrate 100 g/l in aqueous solution
RICC6317-16
:
Ricca Chemical
:
Potassium nitrate 100 g/l in aqueous solution
RICC6317-16
:
Ricca Chemical
:
Pk:
1
Seq:
0001
:
:
101.1 g/mol
:
Ambient
:
231-818-8
:
7757-79-1
:
KNO₃
:
MFCD00011409