5-Bromo-7-nitroindoline ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:80166-90-1
:
Formula:C₈H₇BrN₂O₂
MDL Number:MFCD00005708
:243.06 g/mol
Melting Pt:128…133 °C
Storage Temperature:Ambient
EINECS:279-411-4
UN:2811
ADR:6.1,III
:GHS06
:GHS07