Order Entry
Singapore
ContactUsLinkComponent
Hydrochloric acid 0.05 mol/l (0.05 N)
Hydrochloric acid 0.05 mol/l (0.05 N)
Catalog # RICC3597-1
Supplier:  Ricca Chemical
CAS Number:  
Hydrochloric acid 0.05 mol/l (0.05 N)
Catalog # RICC3597-1
Supplier:  Ricca Chemical
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
Density:1,004 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-595-7
ADR:8,III
 :36.46
UN:1789
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324