

Sodium iodide, granular, AR®, Macron Fine Chemicals™
Catalog # MACR1141-12
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Glass bottle, amber, wide mouth
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 0.02 %
- Alkalinity (as NaOH):≤0.02 %
- Aluminum (Al):≤0.0005 %
- Barium (Ba):≤0.002 %
- Chloride and Bromide (as Cl):≤0.01 %
- Heavy Metals (as Pb):≤0.0005 %
- Insoluble Matter:≤0.005 %
- Iodate (IO₃):≤0.0003 %
- Iron (Fe):≤5 ppm
- Lead (Pb), For Information Only:= 0.001 %
- Nitrogen Compounds (as N):≤0.001 %
- Phosphate (PO₄):≤0.001 %
- Potassium (K):≤0.01 %
- Sulfate (SO₄):≤0.005 %
- Solubility:Passes Test
- Hydrazine Monohydrate:≤0.5 ppm
Specification Test Results
About this item
:
:149.89 g/mol
Merck Index:13,08705
Boiling Pt:1300 °C (1013 hPa)
Melting Pt:662 °C
Density:3,67 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-679-3
UN:3077
ADR:9,III
:GHS09
:GHS07
CAS Number:7681-82-5
Formula:NaI
MDL Number:MFCD00003532