Resorcinol
Glass bottle for solids, 20 mg
Catalog # EDQMY0000767
Supplier: EDQM
Jump to:
Product Details & Documents
Synonyms: 1,3-Dihydroxybenzene, 1,3-Benzenediol
- CAS Number:108-46-3
- Formula:C₆H₆O₂
- MDL Number:MFCD00002269
- Molecular Weight:110.11 g/mol
- Merck Index:12,08323
- Boiling Pt:281 °C (1013 hPa)
- Melting Pt:109…111 °C
- Density:1,272 g/cm³ (20 °C)
- Flash Pt:127 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:203-585-2
- UN:2876
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:20 mg



