Order Entry
Singapore
ContactUsLinkComponent
Magnetic separation devices
Catalog # OMEGMSD-01
Supplier:  OMEGA BIO-TEK
Magnetic separation devices
Catalog # OMEGMSD-01
Supplier:  OMEGA BIO-TEK
Specifications
  • Description:
    E-Z 96 Magnetic separation device-A, fits 96-well microplates
  • Pk:
    1
Specifications
About this item
A range of magnetic devices for separation of proteins or peptides 'bound' to magnetic particles or beads. Magnetic separation is a simple, scaleable and gentle technique that is suitable for crude samples, even those containing particulates.
  • Devices available for 96-well plates and microtubes
  • Simple and effective immobilastion of particles whilst the buffer/elutant is exchanged