9-Fluorenylmethyl chloroformate (Fmoc chloride) ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: (9H-Fluoren-9-yl)methyl carbonochloridate, 9-Fluorenylmethyl chloroformate, Fmoc-Cl, 9H-Fluoren-9-ylmethyl chloroformate, Chloroformic acid 9-fluorenylmethyl ester, FMOCCl
CAS Number:28920-43-6
:
Formula:C₁₅H₁₁ClO₂
MDL Number:MFCD00001138
Molecular Weight:258.7 g/mol
Melting Pt:60…64 °C
EINECS:249-313-6
UN:2923
ADR:8 II
:GHS05
:GHS07
Specifications
- Environmentally Preferable: