
Curcumin 95% (total curcuminoid content)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:458-37-7
:
Formula:C₂₁H₂₀O₆
MDL Number:MFCD00008365
:368.39
Merck Index:14,2673
Melting Pt:170…175 °C
EINECS:207-280-5
:GHS07
Storage Temperature:Ambient