
3-Methyl-1H-pyrazole ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1453-58-3
:
Formula:C₄H₆N₂
MDL Number:MFCD00005240
:82.11 g/mol
Boiling Pt:203…205 °C
Density:1,020
Flash Pt:93 °C (199 °F)
EINECS:215-925-7
:GHS07
Storage Temperature:Ambient