Order Entry
Singapore
ContactUsLinkComponent
 
Water with 0,1% (v/v) formic acid 0,1% (v/v) in water, CHROMASOLV™, Solvent Blend for LC-MS, Riedel-de Haën™
  HONC576913-4X4L
 :  
Water with 0,1% (v/v) formic acid 0,1% (v/v) in water, CHROMASOLV™, Solvent Blend for LC-MS, Riedel-de Haën™
  HONC576913-4X4L
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    16 L
  • Seq:
    5254

 

 

  • Contains:
    0.1 %(v/v) formic acid

 

 

 :
 :0,998 g/cm³ (20 °C)
 :Ambient
 :231-791-2
 :01-2119491174-37-XXXX
 :7732-18-5