Order Entry
Singapore
ContactUsLinkComponent
Accessories for glass thermometers
  BOHLB286-03
 :  Bohlender
undefined
Accessories for glass thermometers
  BOHLB286-03
 :  Bohlender
  • Description:
    Holder (PTFE) for thermometers of Ø 7 - 8 mm, with integrated nut for easy locking/unlocking of the ground joint (NS 29/32)
  • Pk:
    1
Thermometer holder made of PTFE/silicone.
  • Holds thermometers with an Ø of 7 to 8 mm
Features integrated special nut (Safe-Lab) for easy locking and unlocking of the ground joint.