
Bismuth(III) chloride, anhydrous ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7787-60-2
:
Formula:BiCl₃
MDL Number:MFCD00003461
:315.34 g/mol
Merck Index:14,1262
Boiling Pt:∼430 °C (subl.)
Melting Pt:230…232 °C
Density:4,75
Storage Temperature:Ambient
EINECS:232-123-2
UN:3260
ADR:8,III
:GHS05