
Silicon(IV) oxide, amorphous S.A. 175-225 m²/g, fumed
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7631-86-9
:
Formula:SiO₂
MDL Number:MFCD00011232
:60.08 g/mol
Merck Index:14,8493
Boiling Pt:2230 °C
Melting Pt:1710 °C
Density:2,2
EINECS:231-545-4
:GHS08
Storage Temperature:Ambient