Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Strontium chloride
Strontium chloride, ultra dry ≥99.995% (metals basis)
Strontium chloride, ultra dry ≥99.995% (metals basis)
:
Thermo Fisher Scientific
Strontium chloride, ultra dry ≥99.995% (metals basis)
:
Thermo Fisher Scientific
:
10476-85-4
:
:
Cl₂Sr
:
MFCD00011249
:
158.53 g/mol
:
14,8840
:
1250 °C
:
873 °C
:
3,052
:
ampouled under argon
:
Ambient
:
233-971-6
:
GHS05
:
GHS07
msds_ALFA035719
Thermo Scientific Acroseal 25 Years