Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Strontium chloride
Strontium chloride, anhydrous ≥99.5% (metals basis)
Strontium chloride, anhydrous ≥99.5% (metals basis)
:
Thermo Fisher Scientific
Strontium chloride, anhydrous ≥99.5% (metals basis)
:
Thermo Fisher Scientific
:
10476-85-4
:
:
Cl₂Sr
:
MFCD00011249
:
158.53 g/mol
:
14,8840
:
1250 °C
:
873 °C
:
3,052
:
Ambient
:
233-971-6
:
GHS05
:
GHS07
msds_ALFA016790
Thermo Scientific Acroseal 25 Years