Reducing tubing connectors, straight, BOLA
Straight fitting, made of PTFE, with two connectors for elastic tubing with different inner diameters.
- Universal chemical resistance
- Maximum temperature up to 250 °C
- Vacuum: suitable
Straight fitting, made of PTFE, with two connectors for elastic tubing with different inner diameters.