

Rubidium nitrate 99.975% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13126-12-0
:
Formula:NO₃Rb
MDL Number:MFCD00011193
:147.47 g/mol
Boiling Pt:578 °C (1013 hPa)
Melting Pt:310 °C
Density:3,11
Storage Temperature:Ambient
EINECS:236-060-1
UN:1477
ADR:5.1,III
:GHS03
:GHS07