

Sodium nitrate ≥99.5%, Puriss. p.a. ACS, ISO, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7631-99-4
:
Formula:NaNO₃
MDL Number:MFCD00011119
:84.99 g/mol
Merck Index:13,08720
Boiling Pt: Dec. (on heating)
Melting Pt:308 °C
Density:2,26 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-554-3
UN:1498
ADR:5.1,III
REACH:01-2119488221-41
:GHS07
:GHS03
Specification Test Results
- Assay:Min. 99.5 %
- Water insoluble matter:Max. 0.005 %
- pH (5 %, 25 °C):5.5 - 8.0
- Ammonium (NH₄):Max. 0.002 %
- Calcium (Ca):Max. 0.002 %
- Cadmium (Cd):Max. 0.0005 %
- Copper (Cu):Max. 0.0005 %
- Iron (Fe):Max. 0.0002 %
- Potassium (K):Max. 0.005 %
- Magnesium (Mg):Max. 0.001 %
- Lead (Pb):Max. 0.0005 %
- Zinc (Zn):Max. 0.0005 %
- Heavy metals (as Pb):Max. 0.0005 %
- Chloride (Cl):Max. 0.0005 %
- Iodate (IO₃):Max. 0.0005 %
- Nitrite (NO₂):Max. 0.001 %
- Phosphate (PO₄):Max. 0.0002 %
- Sulfate (SO₄):Max. 0.003 %