Sodium nitrate, high purity
VWRC0598-500G
:
- Pack type:HDPE bottle
- Pk:500 g
- Seq:3645
- Insolubles:≤0.04%
- Purity:≥99.0%
:
:84.99 g/mol
:13,08720
:380 °C (1013 hPa)
:308 °C
:2,26 g/cm³ (20 °C)
:Ambient
:231-554-3
:1498
:5.1,III
:01-2119488221-41
:GHS03
:GHS07
:7631-99-4
:NaNO₃
:MFCD00011119