Potassium chloride, ultra dry ≥99.998% (metals basis)
Catalog # ALFA13682.18
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:74.55 g/mol
Merck Index:14,7621
Boiling Pt:1500 °C (subl.)
Melting Pt:773 °C
Density:1,984
:ampouled under argon
Storage Temperature:Ambient
EINECS:231-211-8
CAS Number:7447-40-7
Formula:KCl
MDL Number:MFCD00011360