
Lithium carbonate ≥99.0% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:554-13-2
:
Formula:Li₂CO₃
MDL Number:MFCD00011084
:73.89 g/mol
Merck Index:14,5527
Boiling Pt:1310 °C (dec.)
Melting Pt:723 °C
Density:2,11
EINECS:209-062-5
:GHS07
Storage Temperature:Ambient