Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lithium tetrafluoroborate
Lithium tetrafluoroborate ≥98%
Lithium tetrafluoroborate ≥98%
:
Thermo Fisher Scientific
Lithium tetrafluoroborate ≥98%
:
Thermo Fisher Scientific
:
14283-07-9
:
:
BF₄Li
:
MFCD00011087
:
93.74 g/mol
:
14,5543
:
293…300 °C (dec.)
:
0,852 g/cm³ (20 °C)
:
Ambient
:
238-178-9
:
3260
:
8,III
:
GHS05
:
GHS07
msds_ALFA011528
Thermo Scientific Acroseal 25 Years